Assay Details
Gene Symbol: stimate | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005033.2 | NP_001005033.2 | 1 - 2 | 271 | 64 | |
GenBank mRNA | BC076883.1 | - | 1 - 2 | 221 | 64 | |
CR848301.2 | - | 1 - 2 | 219 | 64 | ||
CX344679.2 | - | 1 - 2 | 271 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
