Assay Details
Gene Symbol: stimate | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005033.2 | NP_001005033.2 | 2 - 3 | 314 | 72 | |
GenBank mRNA | BC076883.1 | - | 2 - 3 | 264 | 72 | |
CR848301.2 | - | 2 - 3 | 262 | 72 | ||
CX344679.2 | - | 2 - 3 | 314 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
