Assay Details
Gene Symbol: stimate | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005033.2 | NP_001005033.2 | 4 - 5 | 538 | 60 | |
GenBank mRNA | BC076883.1 | - | 4 - 5 | 488 | 60 | |
CR848301.2 | - | 4 - 5 | 486 | 60 | ||
CX344679.2 | - | 4 - 5 | 538 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
