Assay Details
Gene Symbol: stimate | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005033.2 | NP_001005033.2 | 5 - 6 | 651 | 89 | |
GenBank mRNA | BC076883.1 | - | 5 - 6 | 601 | 90 | |
CR848301.2 | - | 5 - 6 | 599 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
