Assay Details
| Gene Symbol: rasl10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005037.1 | NP_001005037.1 | 2 - 3 | 388 | 67 | |
| XM_012960002.2 | XP_012815456.1 | 2 - 3 | 648 | 67 | ||
| XM_012960006.2 | XP_012815460.1 | 2 - 3 | 601 | 67 | ||
| GenBank mRNA | BC076894.1 | - | 2 - 3 | 388 | 67 | |
More Information
Set Membership: |
|

rasl10a
Genomic Map