Assay Details
| Gene Symbol: apcdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005044.1 | NP_001005044.1 | 2 - 3 | 432 | 62 | |
| GenBank mRNA | BC076909.1 | - | 2 - 3 | 432 | 62 | |
More Information
Set Membership: |
|

apcdd1
Genomic Map