Assay Details
| Gene Symbol: traf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005074.1 | NP_001005074.1 | 6 - 7 | 1008 | 77 | |
| XM_018091173.1 | XP_017946662.1 | 6 - 7 | 2657 | 77 | ||
| GenBank mRNA | BC076992.1 | - | 6 - 7 | 1008 | 77 | |
| BC168067.1 | - | 1 - 2 | 111 | 77 | ||
More Information
Set Membership: |
|

traf4
Genomic Map