Assay Details
| Gene Symbol: rpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005078.2 | NP_001005078.1 | 2 - 3 | 111 | 91 | |
| XM_012960135.1 | XP_012815589.1 | 3 - 4 | 397 | 91 | ||
| GenBank mRNA | BC077000.1 | - | 2 - 3 | 114 | 91 | |
| CT025372.2 | - | 2 - 3 | 91 | 91 | ||
More Information
Set Membership: |
|

rpl17
Genomic Map