Assay Details
| Gene Symbol: rpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005078.2 | NP_001005078.1 | 4 - 5 | 344 | 85 | |
| XM_012960135.1 | XP_012815589.1 | 5 - 6 | 630 | 85 | ||
| GenBank mRNA | BC077000.1 | - | 4 - 5 | 347 | 85 | |
| CT025372.2 | - | 4 - 5 | 324 | 85 | ||
More Information
Set Membership: |
|

rpl17
Genomic Map