Assay Details
Gene Symbol: ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005080.1 | NP_001005080.1 | 1 - 2 | 276 | 68 | |
GenBank mRNA | BC077002.1 | - | 1 - 2 | 276 | 68 | |
CR761339.2 | - | 1 - 2 | 141 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
