Assay Details
| Gene Symbol: ssr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005080.1 | NP_001005080.1 | 3 - 4 | 493 | 87 | |
| GenBank mRNA | BC077002.1 | - | 3 - 4 | 493 | 87 | |
| CR761339.2 | - | 3 - 4 | 358 | 87 | ||
More Information
Set Membership: |
|

ssr3
Genomic Map