Assay Details
Gene Symbol: csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005081.2 | NP_001005081.1 | 4 - 5 | 405 | 58 | |
XM_018096223.1 | XP_017951712.1 | 4 - 5 | 591 | 58 | ||
GenBank mRNA | BC077003.1 | - | 4 - 5 | 406 | 58 | |
CR762325.2 | - | 3 - 4 | 316 | 58 | ||
CR848278.2 | - | 4 - 5 | 408 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
