Assay Details
Gene Symbol: tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005091.1 | NP_001005091.1 | 3 - 4 | 267 | 99 | |
GenBank mRNA | BC077019.1 | - | 3 - 4 | 267 | 99 | |
CT025310.2 | - | 2 - 3 | 405 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
