Assay Details
Gene Symbol: acot13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005098.2 | NP_001005098.1 | 2 - 3 | 414 | 94 | |
GenBank mRNA | BC077030.1 | - | 2 - 3 | 417 | 94 | |
CT025373.2 | - | 1 - 2 | 200 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
