Assay Details
| Gene Symbol: rpl36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005100.2 | NP_001005100.1 | 2 - 3 | 142 | 73 | |
| GenBank mRNA | BC077033.1 | - | 2 - 3 | 143 | 73 | |
More Information
Set Membership: |
|

rpl36
Genomic Map