Assay Details
Gene Symbol: dnaja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011012.1 | NP_001011012.1 | 8 - 9 | 1107 | 78 | |
GenBank mRNA | BC082725.1 | - | 8 - 9 | 1107 | 78 | |
CT025404.2 | - | 8 - 9 | 1045 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
