Assay Details
| Gene Symbol: dpy30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011017.1 | NP_001011017.1 | 3 - 4 | 267 | 65 | |
| GenBank mRNA | BC082731.1 | - | 3 - 4 | 267 | 65 | |
| CR760705.1 | - | 2 - 3 | 112 | 65 | ||
More Information
Set Membership: |
|

dpy30
Genomic Map