Assay Details
| Gene Symbol: rsrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011036.1 | NP_001011036.1 | 4 - 5 | 530 | 81 | |
| XM_012960614.2 | XP_012816068.1 | 4 - 5 | 557 | 81 | ||
| XM_012960619.2 | XP_012816073.1 | 4 - 5 | 557 | 81 | ||
| XM_012960622.2 | XP_012816076.1 | 4 - 5 | 503 | 81 | ||
| XM_012960627.2 | XP_012816081.1 | 5 - 6 | 581 | 81 | ||
| XM_018092682.1 | XP_017948171.1 | 4 - 5 | 503 | 81 | ||
| GenBank mRNA | BC084062.1 | - | 4 - 5 | 530 | 81 | |
| CR760777.2 | - | 4 - 5 | 520 | 81 | ||
More Information
Set Membership: |
|

rsrc2
Genomic Map