Assay Details
Gene Symbol: slc30a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011041.1 | NP_001011041.1 | 4 - 5 | 679 | 61 | |
XM_018094625.1 | XP_017950114.1 | 3 - 4 | 688 | 61 | ||
GenBank mRNA | BC084148.1 | - | 4 - 5 | 679 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
