Assay Details
Gene Symbol: crhr1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011042.1 | NP_001011042.1 | 12 - 13 | 1313 | 79 | |
XM_012971695.2 | XP_012827149.1 | 12 - 13 | 1389 | 79 | ||
XM_012971696.2 | XP_012827150.1 | 12 - 13 | 1456 | 79 | ||
XM_012971697.2 | XP_012827151.1 | 12 - 13 | 1335 | 79 | ||
GenBank mRNA | BC084149.1 | - | 12 - 13 | 1313 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
