Assay Details
Gene Symbol: crhr1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011042.1 | NP_001011042.1 | 1 - 2 | 128 | 93 | |
XM_012971695.2 | XP_012827149.1 | 1 - 2 | 240 | 93 | ||
XM_012971697.2 | XP_012827151.1 | 1 - 2 | 160 | |||
GenBank mRNA | BC084149.1 | - | 1 - 2 | 128 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012971697.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
