Assay Details
Gene Symbol: timm23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011053.1 | NP_001011053.1 | 3 - 4 | 316 | 81 | |
XM_018095361.1 | XP_017950850.1 | 3 - 4 | 254 | 81 | ||
GenBank mRNA | BC084178.1 | - | 3 - 4 | 316 | 81 | |
CT485778.1 | - | 3 - 4 | 327 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
