Assay Details
| Gene Symbol: zp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203523.1 | NP_988854.1 | 6 - 7 | 844 | 95 | |
| GenBank mRNA | AY079192.1 | - | 6 - 7 | 844 | 95 | |
| CR926385.2 | - | 6 - 7 | 833 | 95 | ||
More Information
Set Membership: |
|

zp4
Genomic Map