Assay Details
| Gene Symbol: vegt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203527.1 | NP_988858.1 | 2 - 3 | 227 | 84 | |
| XM_012958010.1 | XP_012813464.1 | 2 - 3 | 238 | 84 | ||
| GenBank mRNA | AF180352.1 | - | 2 - 3 | 227 | 84 | |
| BC167867.1 | - | 2 - 3 | 240 | 84 | ||
| CR760687.2 | - | 2 - 3 | 302 | 84 | ||
More Information
Set Membership: |
|

vegt
Genomic Map