Assay Details
Gene Symbol: hbg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203528.2 | NP_988859.1 | 1 - 2 | 135 | 106 | |
GenBank mRNA | BC088022.1 | - | 1 - 2 | 111 | 106 | |
M32457.1 | - | 1 - 2 | 137 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
