Assay Details
Gene Symbol: chchd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203547.1 | NP_988878.1 | 2 - 3 | 175 | 73 | |
GenBank mRNA | BC059772.1 | - | 2 - 3 | 175 | 73 | |
CR855780.2 | - | 2 - 3 | 123 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
