Assay Details
| Gene Symbol: myl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203623.1 | NP_988954.1 | 6 - 7 | 706 | 65 | |
| XM_012970239.2 | XP_012825693.1 | 6 - 7 | 829 | 65 | ||
| GenBank mRNA | BC061302.1 | - | 6 - 7 | 706 | 65 | |
| BC089727.1 | - | 6 - 7 | 692 | 65 | ||
| BC169133.1 | - | 689 | 65 | |||
| CR760864.2 | - | 6 - 7 | 675 | 65 | ||
| CR926445.2 | - | 6 - 7 | 505 | 65 | ||
More Information
Set Membership: |
|

myl1
Genomic Map