Assay Details
Gene Symbol: ube2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203625.1 | NP_988956.1 | 2 - 3 | 348 | 103 | |
GenBank mRNA | BC061289.1 | - | 2 - 3 | 348 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
