Assay Details
Gene Symbol: dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203667.1 | NP_988998.1 | 170 | 62 | ||
GenBank mRNA | BC061422.1 | - | 170 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
