Assay Details
Gene Symbol: spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203672.1 | NP_989003.1 | 8 - 9 | 993 | 71 | |
XM_012971593.2 | XP_012827047.1 | 8 - 9 | 1227 | 71 | ||
XM_012971594.2 | XP_012827048.1 | 8 - 9 | 1027 | 71 | ||
XM_012971596.2 | XP_012827050.1 | 8 - 9 | 1005 | 71 | ||
XM_018097685.1 | XP_017953174.1 | 8 - 9 | 1053 | 71 | ||
GenBank mRNA | BC061316.1 | - | 8 - 9 | 993 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
