Assay Details
Gene Symbol: elof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203680.1 | NP_989011.1 | 2 - 3 | 196 | 82 | |
XM_018092000.1 | XP_017947489.1 | 2 - 3 | 239 | 82 | ||
GenBank mRNA | BC061318.1 | - | 2 - 3 | 196 | 82 | |
CR855755.2 | - | 1 - 2 | 143 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
