Assay Details
Gene Symbol: isy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203691.1 | NP_989022.1 | 388 | 63 | ||
GenBank mRNA | BC061420.1 | - | 388 | 63 | ||
CR761820.2 | - | 128 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -




