Assay Details
| Gene Symbol: ube2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203701.1 | NP_989032.1 | 3 - 4 | 664 | 96 | |
| GenBank mRNA | BC061394.1 | - | 3 - 4 | 664 | 96 | |
| CR761582.2 | - | 3 - 4 | 365 | 96 | ||
More Information
Set Membership: |
|

ube2e1
Genomic Map