Assay Details
Gene Symbol: etf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203704.1 | NP_989035.1 | 10 - 11 | 1377 | 70 | |
GenBank mRNA | BC061387.1 | - | 10 - 11 | 1377 | 70 | |
CR762244.2 | - | 9 - 10 | 1234 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
