Assay Details
| Gene Symbol: thap12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203722.1 | NP_989053.1 | 3 - 4 | 660 | 75 | |
| GenBank mRNA | BC061337.1 | - | 3 - 4 | 660 | 75 | |
More Information
Set Membership: |
|

thap12
Genomic Map