Assay Details
| Gene Symbol: rplp0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203736.1 | NP_989067.1 | 6 - 7 | 857 | 72 | |
| XM_012958250.2 | XP_012813704.1 | 5 - 6 | 932 | 72 | ||
| XM_012958254.2 | XP_012813708.1 | 7 - 8 | 952 | 72 | ||
| XM_012958260.1 | XP_012813714.1 | 7 - 8 | 882 | 72 | ||
| XM_012958267.2 | XP_012813721.1 | 6 - 7 | 767 | 72 | ||
| GenBank mRNA | BC061299.1 | - | 6 - 7 | 857 | 72 | |
More Information
Set Membership: |
|

rplp0
Genomic Map