Assay Details
| Gene Symbol: rpsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203737.1 | NP_989068.1 | 5 - 6 | 683 | 65 | |
| GenBank mRNA | BC061298.1 | - | 5 - 6 | 683 | 65 | |
| BC168799.1 | - | 4 - 5 | 659 | 65 | ||
| CR942700.2 | - | 5 - 6 | 673 | 65 | ||
More Information
Set Membership: |
|

rpsa
Genomic Map