Assay Details
Gene Symbol: mdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203743.1 | NP_989074.1 | 4 - 5 | 532 | 65 | |
GenBank mRNA | BC061275.1 | - | 4 - 5 | 532 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
