Assay Details
Gene Symbol: atp1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203756.1 | NP_989087.1 | 1 - 2 | 203 | 65 | |
GenBank mRNA | BC062517.1 | - | 1 - 2 | 203 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
