Assay Details
Gene Symbol: atp5pb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203787.1 | NP_989118.1 | 4 - 5 | 407 | 64 | |
GenBank mRNA | BC061296.1 | - | 4 - 5 | 407 | 64 | |
CR760878.2 | - | 4 - 5 | 439 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
