Assay Details
| Gene Symbol: rps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203788.1 | NP_989119.1 | 5 - 6 | 575 | 78 | |
| GenBank mRNA | BC061265.1 | - | 5 - 6 | 575 | 78 | |
| CR848247.2 | - | 5 - 6 | 546 | 78 | ||
More Information
Set Membership: |
|

rps3
Genomic Map