Assay Details
Gene Symbol: ddx19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203796.1 | NP_989127.1 | 10 - 11 | 1417 | 71 | |
GenBank mRNA | BC061342.1 | - | 10 - 11 | 1417 | 71 | |
CR760667.2 | - | 10 - 11 | 1404 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
