Assay Details
Gene Symbol: ddx19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203796.1 | NP_989127.1 | 4 - 5 | 433 | 70 | |
GenBank mRNA | BC061342.1 | - | 4 - 5 | 433 | 70 | |
CR760667.2 | - | 4 - 5 | 420 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
