Assay Details
| Gene Symbol: tmem30a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203802.1 | NP_989133.1 | 6 - 7 | 910 | 65 | |
| XM_018093822.1 | XP_017949311.1 | 5 - 6 | 917 | 65 | ||
| GenBank mRNA | BC061349.1 | - | 6 - 7 | 910 | 65 | |
| CR761943.2 | - | 6 - 7 | 958 | 65 | ||
More Information
Set Membership: |
|

tmem30a
Genomic Map