Assay Details
| Gene Symbol: rps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203821.1 | NP_989152.1 | 3 - 4 | 373 | 71 | |
| GenBank mRNA | BC061628.1 | - | 3 - 4 | 373 | 71 | |
| BC082345.1 | - | 3 - 4 | 366 | 71 | ||
| BC161304.1 | - | 366 | 71 | |||
| CR760876.2 | - | 3 - 4 | 360 | 71 | ||
| Gene Symbol: tmed2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC169186.1 | - | 1587 | 71 | ||
More Information
Set Membership: |
|

rps6
Genomic Map