Assay Details
| Gene Symbol: etfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203823.1 | NP_989154.1 | 3 - 4 | 443 | 64 | |
| GenBank mRNA | BC061629.1 | - | 3 - 4 | 443 | 64 | |
| BC080971.1 | - | 3 - 4 | 443 | 64 | ||
| CR942746.2 | - | 3 - 4 | 438 | 64 | ||
More Information
Set Membership: |
|

etfb
Genomic Map