Assay Details
| Gene Symbol: gfod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203839.1 | NP_989170.1 | 1 - 2 | 395 | 71 | |
| GenBank mRNA | BC063342.1 | - | 1 - 2 | 395 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gfod1| Gene Symbol: gfod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203839.1 | NP_989170.1 | 1 - 2 | 395 | 71 | |
| GenBank mRNA | BC063342.1 | - | 1 - 2 | 395 | 71 | |
Set Membership: |
|