Assay Details
| Gene Symbol: emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203850.1 | NP_989181.1 | 5 - 6 | 673 | 66 | |
| GenBank mRNA | BC063338.1 | - | 5 - 6 | 673 | 66 | |
| CR760835.2 | - | 5 - 6 | 662 | 66 | ||
More Information
Set Membership: |
|

emc8
Genomic Map