Assay Details
Gene Symbol: bax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203854.1 | NP_989185.1 | 3 - 4 | 328 | 59 | |
GenBank mRNA | BC063201.1 | - | 3 - 4 | 328 | 59 | |
CR855728.2 | - | 3 - 4 | 381 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
