Assay Details
Gene Symbol: cd82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203860.1 | NP_989191.1 | 2 - 3 | 305 | 103 | |
XM_012960761.2 | XP_012816215.1 | 2 - 3 | 157 | 103 | ||
GenBank mRNA | BC063219.1 | - | 2 - 3 | 305 | 103 | |
CR760215.2 | - | 2 - 3 | 274 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
